Assay Details
Gene Symbol: NUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126895.4 | NP_178939.2 | 5 - 6 | 815 | 72 | |
GenBank mRNA | BT022114.1 | - | 5 - 6 | 743 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
