Assay Details
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337542.1 | NP_001327439.1 | 14 - 15 | 1822 | 94 | |
NM_111353.4 | NP_187132.1 | 15 - 16 | 1470 | 94 | ||
NM_202492.3 | NP_974221.2 | 14 - 15 | 1462 | 94 | ||
GenBank mRNA | BT000875.1 | - | 13 - 14 | 1148 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
