Assay Details
Gene Symbol: AT4G29400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119085.4 | NP_194669.2 | 6 - 7 | 887 | 78 | |
GenBank mRNA | AY735656.1 | - | 6 - 7 | 856 | 78 | |
AY924824.1 | - | 6 - 7 | 804 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
