Assay Details
Gene Symbol: PLDDELTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119745.3 | NP_567989.1 | 4 - 5 | 1887 | 86 | |
NM_179170.3 | NP_849501.1 | 4 - 5 | 2091 | 86 | ||
NM_179171.2 | NP_849502.1 | 4 - 5 | 1854 | 86 | ||
GenBank mRNA | AB031047.1 | - | 4 - 5 | 1701 | 86 | |
AF306345.1 | - | 4 - 5 | 1565 | 86 | ||
AF322228.1 | - | 4 - 5 | 1574 | 86 | ||
AK227213.1 | - | 4 - 5 | 1697 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
