Assay Details
Gene Symbol: PLDDELTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119745.3 | NP_567989.1 | 7 - 8 | 2255 | 85 | |
NM_179170.3 | NP_849501.1 | 7 - 8 | 2459 | 85 | ||
NM_179171.2 | NP_849502.1 | 7 - 8 | 2222 | 85 | ||
GenBank mRNA | AB031047.1 | - | 7 - 8 | 2069 | 85 | |
AF306345.1 | - | 7 - 8 | 1933 | 85 | ||
AF322228.1 | - | 7 - 8 | 1942 | 85 | ||
AK227213.1 | - | 7 - 8 | 2065 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
