Assay Details
Gene Symbol: THA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035924.2 | NP_001031001.2 | 7 - 8 | 995 | 127 | |
NM_001084023.1 | NP_001077492.1 | 7 - 8 | 993 | 85 | ||
NM_001331780.1 | NP_001318956.1 | 6 - 7 | 1006 | 127 | ||
NM_001331781.1 | NP_001322023.1 | 7 - 8 | 956 | 127 | ||
NM_100736.3 | NP_563822.1 | 7 - 8 | 1472 | 127 | ||
NM_179283.2 | NP_849614.1 | 7 - 8 | 1035 | 127 | ||
NM_179284.4 | NP_849615.1 | 7 - 8 | 1068 | 127 | ||
GenBank mRNA | AK230078.1 | - | 7 - 8 | 1077 | 127 | |
AK317034.1 | - | 7 - 8 | 958 | 85 | ||
AY080670.1 | - | 7 - 8 | 1031 | 127 | ||
AY117227.1 | - | 5 - 6 | 866 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
