Assay Details
Gene Symbol: MYB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112118.3 | NP_187888.1 | 1 - 2 | 161 | 66 | |
GenBank mRNA | AY519586.1 | - | 1 - 2 | 132 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
