Assay Details
Gene Symbol: MYB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338009.1 | NP_001325605.1 | 1 - 2 | 211 | 81 | |
NM_112118.3 | NP_187888.1 | 2 - 3 | 295 | 81 | ||
GenBank mRNA | AY519586.1 | - | 2 - 3 | 266 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
