Assay Details
Gene Symbol: SRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112145.4 | NP_566444.4 | 12 - 13 | 1847 | 65 | |
GenBank mRNA | AY518289.1 | - | 12 - 13 | 1847 | 65 |
Gene Symbol: AT3G13062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ154118.1 | - | 18 - 19 | 3382 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
