Assay Details
Gene Symbol: AT3G13930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112247.3 | NP_566470.1 | 5 - 6 | 583 | 90 | |
GenBank mRNA | AF367302.1 | - | 5 - 6 | 413 | 90 | |
AY091691.1 | - | 4 - 5 | 273 | 90 | ||
BT000444.1 | - | 5 - 6 | 413 | 90 | ||
BT000702.1 | - | 5 - 6 | 415 | 90 | ||
BT001223.1 | - | 4 - 5 | 273 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
