Assay Details
Gene Symbol: AT3G14910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112353.4 | NP_188109.1 | 3 - 4 | 693 | 70 | |
GenBank mRNA | BT022104.1 | - | 3 - 4 | 678 | 70 | |
BT029181.1 | - | 3 - 4 | 672 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
