Assay Details
Gene Symbol: AT3G14910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112353.4 | NP_188109.1 | 4 - 5 | 753 | 73 | |
GenBank mRNA | BT022104.1 | - | 4 - 5 | 738 | 73 | |
BT029181.1 | - | 4 - 5 | 732 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
