Assay Details
Gene Symbol: CCD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119434.4 | NP_195007.2 | 4 - 5 | 1586 | 68 | |
GenBank mRNA | AY074264.1 | - | 4 - 5 | 1582 | 68 | |
AY133732.1 | - | 4 - 5 | 1466 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
