Assay Details
Gene Symbol: CCD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342177.1 | NP_001329787.1 | 4 - 5 | 1763 | 70 | |
NM_119434.4 | NP_195007.2 | 5 - 6 | 1685 | 70 | ||
GenBank mRNA | AY074264.1 | - | 5 - 6 | 1681 | 70 | |
AY133732.1 | - | 5 - 6 | 1565 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
