Assay Details
Gene Symbol: EDA39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125635.2 | NP_001119107.1 | 7 - 8 | 1375 | 141 | |
NM_001342196.1 | NP_001328068.1 | 8 - 9 | 1336 | 141 | ||
NM_001342197.1 | NP_001328069.1 | 7 - 8 | 1386 | 141 | ||
NM_119459.4 | NP_195031.2 | 6 - 7 | 1489 | 141 | ||
NM_202943.3 | NP_974672.1 | 7 - 8 | 1404 | 141 | ||
NM_202944.3 | NP_974673.2 | 8 - 9 | 1290 | 141 | ||
GenBank mRNA | AY062551.1 | - | 6 - 7 | 1486 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
