Assay Details
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203987.1 | NP_001190916.1 | 3 - 4 | 620 | 119 | |
NM_001203988.2 | NP_001190917.1 | 3 - 4 | 388 | 119 | ||
NM_001342313.1 | NP_001327974.1 | 3 - 4 | 388 | 119 | ||
NM_001342314.1 | NP_001327973.1 | 1 - 2 | 339 | 119 | ||
NM_119668.6 | NP_195228.1 | 1 - 2 | 360 | 119 | ||
GenBank mRNA | AF031427.2 | - | 1 - 2 | 270 | 119 | |
BT020318.1 | - | 1 - 2 | 162 | 119 | ||
BT020603.1 | - | 1 - 2 | 103 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
