Assay Details
Gene Symbol: MSI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119671.3 | NP_195231.1 | 4 - 5 | 1089 | 64 | |
GenBank mRNA | BT020491.1 | - | 4 - 5 | 942 | 64 | |
BT025992.1 | - | 4 - 5 | 912 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
