Assay Details
Gene Symbol: AT2G17760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179636.3 | NP_849967.1 | 4 - 5 | 1156 | 82 | |
GenBank mRNA | AY069886.1 | - | 4 - 5 | 1042 | 82 | |
AY142012.1 | - | 4 - 5 | 902 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
