Assay Details
Gene Symbol: ARF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084486.1 | NP_001077955.1 | 2 - 3 | 243 | 114 | |
NM_179726.3 | NP_850057.1 | 2 - 3 | 281 | 114 | ||
GenBank mRNA | AK117735.1 | - | 2 - 3 | 224 | 114 | |
AY063840.1 | - | 2 - 3 | 243 | 114 | ||
AY150410.1 | - | 2 - 3 | 147 | 114 | ||
X77385.1 | - | 175 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
