Assay Details
Gene Symbol: ARF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084486.1 | NP_001077955.1 | 7 - 8 | 567 | 68 | |
NM_179726.3 | NP_850057.1 | 7 - 8 | 642 | 68 | ||
GenBank mRNA | AK117735.1 | - | 7 - 8 | 585 | 68 | |
AY063840.1 | - | 7 - 8 | 604 | 68 | ||
AY150410.1 | - | 7 - 8 | 508 | 68 | ||
X77385.1 | - | 536 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
