Assay Details
Gene Symbol: AT3G16370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112508.5 | NP_188258.1 | 1 - 2 | 457 | 82 | |
GenBank mRNA | AY075698.1 | - | 1 - 2 | 263 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
