Assay Details
Gene Symbol: AT3G16370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112508.5 | NP_188258.1 | 4 - 5 | 1085 | 149 | |
GenBank mRNA | AY075698.1 | - | 4 - 5 | 891 | 149 | |
BT002609.1 | - | 4 - 5 | 867 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
