Assay Details
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035654.2 | NP_001030731.1 | 3 - 4 | 362 | 154 | |
NM_001338474.1 | NP_001327849.1 | 4 - 5 | 440 | 154 | ||
NM_112920.4 | NP_566657.1 | 4 - 5 | 507 | 154 | ||
GenBank mRNA | AK317308.1 | - | 3 - 4 | 428 | 154 | |
AY034911.1 | - | 4 - 5 | 364 | 154 | ||
BT004414.1 | - | 3 - 4 | 234 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
