Assay Details
Gene Symbol: LTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342367.1 | NP_001329911.1 | 4 - 5 | 808 | 90 | |
NM_119742.4 | NP_567988.1 | 4 - 5 | 808 | 90 | ||
GenBank mRNA | AY099728.1 | - | 4 - 5 | 794 | 90 | |
AY128903.1 | - | 4 - 5 | 753 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
