Assay Details
Gene Symbol: AT4G36440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119807.2 | NP_195362.2 | 6 - 7 | 469 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
