Assay Details
Gene Symbol: HTR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035850.3 | NP_001030927.1 | 3 - 4 | 424 | 72 | |
NM_001331269.1 | NP_001322867.1 | 4 - 5 | 337 | 72 | ||
NM_001331270.1 | NP_001322868.1 | 4 - 5 | 307 | 72 | ||
NM_100019.2 | NP_563627.1 | 4 - 5 | 337 | 72 | ||
GenBank mRNA | AB081500.1 | - | 3 - 4 | 149 | 72 | |
AF465800.1 | - | 3 - 4 | 140 | 72 | ||
AK175840.1 | - | 3 - 4 | 196 | 72 | ||
BT024771.1 | - | 3 - 4 | 140 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
