Assay Details
Gene Symbol: HTR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035850.3 | NP_001030927.1 | 8 - 9 | 767 | 59 | |
NM_001331269.1 | NP_001322867.1 | 9 - 10 | 680 | 59 | ||
NM_001331270.1 | NP_001322868.1 | 9 - 10 | 650 | 59 | ||
NM_100019.2 | NP_563627.1 | 9 - 10 | 680 | 59 | ||
GenBank mRNA | AB081500.1 | - | 8 - 9 | 492 | 59 | |
AF465800.1 | - | 8 - 9 | 483 | 59 | ||
AK175840.1 | - | 8 - 9 | 539 | 59 | ||
BT024771.1 | - | 8 - 9 | 483 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
