Assay Details
Gene Symbol: EDA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100076.3 | NP_171698.1 | 6 - 7 | 3157 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
