Assay Details
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100148.3 | NP_171768.2 | 3 - 4 | 273 | 100 | |
GenBank mRNA | AB493426.1 | - | 2 - 3 | 138 | 100 | |
AK175906.1 | - | 3 - 4 | 273 | 100 | ||
AK175966.1 | - | 3 - 4 | 273 | 100 | ||
AK176854.1 | - | 3 - 4 | 241 | 100 | ||
AK221393.1 | - | 3 - 4 | 252 | 100 | ||
AY463615.1 | - | 2 - 3 | 138 | 100 | ||
BT010482.1 | - | 2 - 3 | 138 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
