Assay Details
Gene Symbol: AT1G05310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100410.3 | NP_172023.1 | 1 - 2 | 537 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
