Assay Details
Gene Symbol: EDA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202850.1 | NP_001189779.1 | 3 - 4 | 441 | 65 | |
NM_130381.3 | NP_566127.1 | 3 - 4 | 522 | 65 | ||
GenBank mRNA | AB246325.1 | - | 3 - 4 | 413 | 65 | |
AY045643.1 | - | 3 - 4 | 446 | 65 | ||
AY059652.1 | - | 3 - 4 | 413 | 65 | ||
AY088054.1 | - | 3 - 4 | 422 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
