Assay Details
Gene Symbol: AT1G09010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100772.5 | NP_563833.2 | 10 - 11 | 2251 | 78 | |
GenBank mRNA | AB122060.1 | - | 10 - 11 | 2092 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
