Assay Details
Gene Symbol: AT1G09010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100772.5 | NP_563833.2 | 2 - 3 | 440 | 84 | |
GenBank mRNA | AB122060.1 | - | 2 - 3 | 281 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
