Assay Details
Gene Symbol: AT1G09010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100772.5 | NP_563833.2 | 3 - 4 | 675 | 66 | |
GenBank mRNA | AB122060.1 | - | 3 - 4 | 516 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
