Assay Details
Gene Symbol: AT1G09390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100809.5 | NP_172410.1 | 3 - 4 | 829 | 84 | |
GenBank mRNA | BT002885.1 | - | 3 - 4 | 647 | 84 | |
BT004397.1 | - | 3 - 4 | 611 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
