Assay Details
Gene Symbol: AT5G57040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125086.4 | NP_200514.1 | 1 - 2 | 178 | 78 | |
GenBank mRNA | AK227223.1 | - | 1 - 2 | 75 | 78 | |
AY088499.1 | - | 1 - 2 | 105 | 78 | ||
BT024587.1 | - | 1 - 2 | 48 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
