Assay Details
Gene Symbol: AT5G57040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125086.4 | NP_200514.1 | 2 - 3 | 351 | 101 | |
GenBank mRNA | AK227223.1 | - | 2 - 3 | 248 | 101 | |
AY088499.1 | - | 2 - 3 | 278 | 101 | ||
BT024587.1 | - | 2 - 3 | 221 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
