Assay Details
Gene Symbol: CER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125164.3 | NP_200588.2 | 6 - 7 | 1532 | 120 | |
GenBank mRNA | AB099512.1 | - | 6 - 7 | 1504 | 120 | |
AB116548.1 | - | 6 - 7 | 1213 | 120 | ||
AY131334.1 | - | 6 - 7 | 1503 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
