Assay Details
Gene Symbol: FIP1[V] | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345286.1 | NP_001332116.1 | 5 - 6 | 1461 | 69 | |
NM_125189.3 | NP_200612.2 | 5 - 6 | 1458 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
