Assay Details
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203645.2 | NP_001190574.1 | 3 - 4 | 869 | 111 | |
NM_001203646.1 | NP_001190575.1 | 3 - 4 | 825 | 111 | ||
NM_001345386.1 | NP_001331790.1 | 2 - 3 | 1342 | 111 | ||
NM_001345387.1 | NP_001331791.1 | 2 - 3 | 1342 | 111 | ||
NM_001345388.1 | NP_001331792.1 | 3 - 4 | 865 | 111 | ||
NM_001345389.1 | NP_001331789.1 | 3 - 4 | 818 | 111 | ||
NM_125403.3 | NP_568919.1 | 3 - 4 | 866 | 111 | ||
GenBank mRNA | AB046956.1 | - | 2 - 3 | 429 | 111 | |
AB493801.1 | - | 2 - 3 | 429 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
