Assay Details
Gene Symbol: ARF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345418.1 | NP_001331788.1 | 12 - 13 | 2724 | 91 | |
NM_125438.4 | NP_200853.1 | 11 - 12 | 2796 | 91 | ||
GenBank mRNA | AF013466.1 | - | 11 - 12 | 2254 | 91 | |
AF344313.1 | - | 11 - 12 | 2199 | 91 | ||
AY080832.1 | - | 11 - 12 | 2556 | 91 | ||
AY113977.1 | - | 11 - 12 | 2199 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
