Assay Details
Gene Symbol: ARF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345418.1 | NP_001331788.1 | 5 - 6 | 1228 | 68 | |
NM_125438.4 | NP_200853.1 | 4 - 5 | 1300 | 68 | ||
GenBank mRNA | AF013466.1 | - | 4 - 5 | 758 | 68 | |
AF344313.1 | - | 4 - 5 | 703 | 68 | ||
AY080832.1 | - | 4 - 5 | 1060 | 68 | ||
AY113977.1 | - | 4 - 5 | 703 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
