Assay Details
Gene Symbol: MIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125539.2 | NP_200954.1 | 10 - 11 | 1394 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
