Assay Details
Gene Symbol: MES5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343099.1 | NP_001332228.1 | 1 - 2 | 425 | 59 | |
NM_121068.4 | NP_196592.1 | 1 - 2 | 587 | 59 | ||
GenBank mRNA | AK226255.1 | - | 1 - 2 | 423 | 59 | |
AY093714.1 | - | 1 - 2 | 358 | 59 | ||
AY142490.1 | - | 1 - 2 | 358 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
