Assay Details
Gene Symbol: AT2G26230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128180.5 | NP_180191.1 | 6 - 7 | 770 | 64 | |
GenBank mRNA | AY050318.1 | - | 6 - 7 | 673 | 64 | |
AY087805.1 | - | 6 - 7 | 723 | 64 | ||
AY101530.1 | - | 5 - 6 | 573 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
