Assay Details
Gene Symbol: TWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113059.3 | NP_188801.2 | 4 - 5 | 449 | 66 | |
GenBank mRNA | AJ224640.1 | - | 4 - 5 | 449 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
