Assay Details
Gene Symbol: NET1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338597.1 | NP_001319620.1 | 1 - 2 | 300 | 134 | |
NM_001338598.1 | NP_001326409.1 | 2 - 3 | 350 | 134 | ||
NM_113178.3 | NP_188918.2 | 2 - 3 | 324 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
