Assay Details
Gene Symbol: MYB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113226.5 | NP_188966.1 | 1 - 2 | 413 | 145 | |
GenBank mRNA | AK176617.1 | - | 1 - 2 | 413 | 145 | |
AK176693.1 | - | 1 - 2 | 413 | 145 | ||
AY550296.1 | - | 1 - 2 | 130 | 145 | ||
BT026046.1 | - | 1 - 2 | 130 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
