Assay Details
Gene Symbol: MSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113339.4 | NP_189075.2 | 5 - 6 | 702 | 85 | |
GenBank mRNA | AY191303.1 | - | 5 - 6 | 668 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
