Assay Details
Gene Symbol: AT5G64100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125806.3 | NP_201215.1 | 2 - 3 | 772 | 72 | |
GenBank mRNA | X98808.1 | - | 642 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
